accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOClN

CAS RN 52092-47-4

IUPAC Name: 5-chloro-2-nitropyridine
Synonyms: 5-chloro-2-nitropyridine
Molecular Weight (g/mol): 158.54
Molecular Formula: C5H3ClN2O2
InChi Key: YUBHMOQVHOODEI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results