CAS RN 521-13-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cholesteryl N-Butyrate, TRC
CAS: 521-13-1 Molecular Formula: C31H52O2 Molecular Weight (g/mol): 456.74 IUPAC Name: [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] butanoate SMILES: CCCC(O[C@H]1CC[C@]2(C(C1)=CC[C@H]3[C@@H]4CC[C@@H]([C@]4(CC[C@@H]32)C)[C@@H](CCCC(C)C)C)C)=O