CAS RN 52179-28-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ciprofibrate Ethyl Ester, TRC
CAS: 52179-28-9 Molecular Formula: C15 H18 Cl2 O3 Molecular Weight (g/mol): 317.21 Synonym: 2-[4-(2,2-dichlorocyclopropyl)phenoxy]-2-methylpropanoic Acid Ethyl Ester,Ciprofibrate ethyl ester,Ethyl 2-[p-(2,2-dichlorocyclopropyl)phenoxy]-2-methylpropionate,Ethyl ciprofibrate,Ethyl 2-[4-[(1RS)-2,2-Dichlorocyclopropyl]phenoxy]-2-methylpropanoate,Ciprofibrate Imp. E (EP) IUPAC Name: ethyl 2-[4-(2,2-dichlorocyclopropyl)phenoxy]-2-methylpropanoate SMILES: CCOC(=O)C(C)(C)Oc1ccc(cc1)C2CC2(Cl)Cl