accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SOOCl

CAS RN 52205-85-3

IUPAC Name: 2,3-dihydro-1H-indene-5-sulfonyl chloride
Synonyms: 2,3-dihydro-1H-indene-5-sulfonyl chloride
Molecular Weight (g/mol): 216.68
Molecular Formula: C9H9ClO2S
InChi Key: SWLIXMXSCZYVTQ-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC2=C(CCC2)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (1)
  • (4)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (5)
  • (5)
  • (1)
  • (4)
  • (5)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)