accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHNN

CAS RN 5221-44-3

IUPAC Name: N-(pyridin-4-yl)benzamide
Synonyms: N-(pyridin-4-yl)benzamide
Molecular Weight (g/mol): 198.23
Molecular Formula: C12H10N2O
InChi Key: PNWVOLKZHJEWQU-UHFFFAOYSA-N
SMILES: O=C(NC1=CC=NC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results