accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 52240-08-1

NHNON

CAS RN 52240-08-1

IUPAC Name: 3-(6-oxo-1,6-dihydropyridazin-3-yl)benzonitrile
Synonyms: 3-(6-oxo-1H-pyridazin-3-yl)benzonitrile
Molecular Weight (g/mol): 197.2
Molecular Formula: C11H7N3O
InChi Key: INFIYPRKBSENAH-UHFFFAOYSA-N
SMILES: O=C1NN=C(C=C1)C1=CC=CC(=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results