accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaOSOOHNNO(E)

CAS RN 523-44-4

IUPAC Name: sodium 4-{2-[(1E)-4-oxo-1,4-dihydronaphthalen-1-ylidene]hydrazin-1-yl}benzene-1-sulfonate
Synonyms: FD&C orange number 1
Molecular Weight (g/mol): 350.32
Molecular Formula: C16H11N2NaO4S
InChi Key: HMWJVUNISIEXFR-FLNCGGNMSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=C(N\N=C2/C=CC(=O)C3=CC=CC=C23)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)