accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3ClClOONO

CAS RN 52315-07-8

IUPAC Name: cyano(3-phenoxyphenyl)methyl 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
Synonyms: ustaad chinmix Colt supercypermethrin ardap ecofleece sheep dip (non-OP) ambush CY β-cypermethrin isomer rycopel neramethrin EC 50
Molecular Weight (g/mol): 416.3
Molecular Formula: C22H19Cl2NO3
InChi Key: KAATUXNTWXVJKI-UHFFFAOYNA-N
SMILES: CC1(C)C(C=C(Cl)Cl)C1C(=O)OC(C#N)C1=CC=CC(OC2=CC=CC=C2)=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
  • (1)
Concentration 
  • (2)
  • (1)
Industry Type 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)