accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFClN

CAS RN 52334-81-3

IUPAC Name: 2-chloro-5-(trifluoromethyl)pyridine
Synonyms: 2-chloro-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 181.54
Molecular Formula: C6H3ClF3N
InChi Key: JFZJMSDDOOAOIV-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(Cl)N=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)