accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 52356-01-1

HClH2NNHOHO

CAS RN 52356-01-1

IUPAC Name: hydrogen 2-hydrazinylbenzoic acid chloride
Synonyms: hydrogen 2-hydrazinylbenzoic acid chloride
Molecular Weight (g/mol): 188.61
Molecular Formula: C7H9ClN2O2
InChi Key: ZGNNOFKURIXXRF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=CC=C1C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results