accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HHClClCH3NOHH2NHO

CAS RN 524-36-7

IUPAC Name: dihydrogen 4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol dichloride
Synonyms: dihydrogen pyridoxamine dichloride
Molecular Weight (g/mol): 241.11
Molecular Formula: C8H14Cl2N2O2
InChi Key: HNWCOANXZNKMLR-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CC1=NC=C(CO)C(CN)=C1O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results