accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 52410-74-9

CH3SOOOOSOOH3C

CAS RN 52410-74-9

IUPAC Name: 2-(methanesulfonyloxy)ethyl methanesulfonate
Synonyms: ethylene dimethanesulfonate
Molecular Weight (g/mol): 218.24
Molecular Formula: C4H10O6S2
InChi Key: QSQFARNGNIZGAW-UHFFFAOYSA-N
SMILES: CS(=O)(=O)OCCOS(C)(=O)=O
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (2)