accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HNNCH3NOCH3N

CAS RN 5248-39-5

IUPAC Name: 4-methoxy-N,6-dimethyl-1,3,5-triazin-2-amine
Synonyms: 4-methoxy-N,6-dimethyl-1,3,5-triazin-2-amine
Molecular Weight (g/mol): 154.17
Molecular Formula: C6H10N4O
InChi Key: MNDSUSQBIDHEJU-UHFFFAOYSA-N
SMILES: CNC1=NC(C)=NC(OC)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results