accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrHN

CAS RN 52488-36-5

IUPAC Name: 4-bromo-1H-indole
Synonyms: 4-bromo-1H-indole
Molecular Weight (g/mol): 196.05
Molecular Formula: C8H6BrN
InChi Key: GRJZJFUBQYULKL-UHFFFAOYSA-N
SMILES: BrC1=C2C=CNC2=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)