accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 525-79-1

NHNNNHNO

CAS RN 525-79-1

IUPAC Name: N-[(furan-2-yl)methyl]-7H-purin-6-amine
Synonyms: 6 furfurylaminopurine kinetin
Molecular Weight (g/mol): 215.22
Molecular Formula: C10H9N5O
InChi Key: QANMHLXAZMSUEX-UHFFFAOYSA-N
SMILES: C(NC1=C2NC=NC2=NC=N1)C1=CC=CO1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)