accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 52507-57-0

IUPAC Name: 7-methoxy-4-methyl-1,2-dihydroquinoline-2-thione
Synonyms: 7-methoxy-4-methyl-1H-quinoline-2-thione
Molecular Weight (g/mol): 205.28
Molecular Formula: C11H11NOS
InChi Key: QJNQZUVUCGOPFS-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(C)=CC(=S)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results