accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaOHOHOOOHO

CAS RN 526-94-3

IUPAC Name: sodium 3-carboxy-2,3-dihydroxypropanoate
Synonyms: monosodium tartrate
Molecular Weight (g/mol): 172.07
Molecular Formula: C4H5NaO6
InChi Key: NKAAEMMYHLFEFN-UHFFFAOYNA-M
SMILES: [Na+].OC(C(O)C([O-])=O)C(O)=O
Percent Purity 
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)