accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5263-02-5

OOOOOOOHOHOHOHOHOHZnZnZnZnZn

CAS RN 5263-02-5

IUPAC Name: pentazinc(2+) hexahydroxide dicarbonate
Synonyms: pentazinc(2+) hexahydroxide dicarbonate
Molecular Weight (g/mol): 548.96
Molecular Formula: C2H6O12Zn5
InChi Key: UOURRHZRLGCVDA-UHFFFAOYSA-D
SMILES: [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Zn++].[Zn++].[Zn++].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)