accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClClClClCl

CAS RN 52663-72-6

IUPAC Name: 2,3',4,4',5,5'-hexachloro-1,1'-biphenyl
Synonyms: 2,3',4,4',5,5'-hexachloro-1,1'-biphenyl
Molecular Weight (g/mol): 360.86
Molecular Formula: C12H4Cl6
InChi Key: AZXHAWRMEPZSSV-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(C=C1Cl)C1=CC(Cl)=C(Cl)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results