accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH2HCl

CAS RN 5267-49-2

IUPAC Name: 1-(4-methylphenyl)-1-phenylmethanamine hydrochloride
Synonyms: 1-(4-methylphenyl)-1-phenylmethanamine hydrochloride
Molecular Weight (g/mol): 233.74
Molecular Formula: C14H16ClN
InChi Key: NHTZSJKMWBONMD-UHFFFAOYNA-N
SMILES: Cl.CC1=CC=C(C=C1)C(N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results