accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaHOOHOHOHOHOO

CAS RN 527-07-1

IUPAC Name: sodium 2,3,4,5,6-pentahydroxyhexanoate
Synonyms: sodium gluconate
Molecular Weight (g/mol): 218.14
Molecular Formula: C6H11NaO7
InChi Key: UPMFZISCCZSDND-UHFFFAOYNA-M
SMILES: [Na+].OCC(O)C(O)C(O)C(O)C([O-])=O
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (2)
  • (3)
  • (7)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (4)
  • (1)