accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 527-21-9

FFOFOF

CAS RN 527-21-9

IUPAC Name: tetrafluorocyclohexa-2,5-diene-1,4-dione
Synonyms: tetrafluorocyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol): 180.06
Molecular Formula: C6F4O2
InChi Key: JKLYZOGJWVAIQS-UHFFFAOYSA-N
SMILES: FC1=C(F)C(=O)C(F)=C(F)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results