accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHH3CH3C

CAS RN 527-54-8

IUPAC Name: 3,4,5-trimethylphenol
Synonyms: 3,4,5-trimethylphenol 3,4,5-hemimellitenol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: FDQQNNZKEJIHMS-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC(C)=C1C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results