accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 527-62-8

NH2ClClHO

CAS RN 527-62-8

IUPAC Name: 2-amino-4,6-dichlorophenol
Synonyms: 2-amino-4,6-dichlorophenol
Molecular Weight (g/mol): 178.01
Molecular Formula: C6H5Cl2NO
InChi Key: WASQBNCGNUTVNI-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC(Cl)=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results