accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONONHN

CAS RN 527-73-1

IUPAC Name: 2-nitro-1H-imidazole
Synonyms: 2-nitroimidazole
Molecular Weight (g/mol): 113.08
Molecular Formula: C3H3N3O2
InChi Key: YZEUHQHUFTYLPH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=NC=CN1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results