accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CN

CAS RN 52709-83-8

IUPAC Name: 4'-butyl-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-butyl-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 235.33
Molecular Formula: C17H17N
InChi Key: PJPLBHHDTUICNN-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results