accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClH3COONO

CAS RN 52763-21-0

IUPAC Name: hydrogen ethyl 1-benzyl-3-oxopiperidine-4-carboxylate chloride
Synonyms: hydrogen ethyl 1-benzyl-3-oxopiperidine-4-carboxylate chloride
Molecular Weight (g/mol): 297.78
Molecular Formula: C15H20ClNO3
InChi Key: UQOMEAWPKSISII-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCOC(=O)C1CCN(CC2=CC=CC=C2)CC1=O
Grade 
  • (5)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results