CAS RN 528-45-0
IUPAC Name:
3,4-dinitrobenzoic acid
Synonyms:
3,4-dinitrobenzoic acid
Molecular Weight (g/mol):
212.12
Molecular Formula:
C7H4N2O6
InChi Key:
OMVRRHJJQILIJX-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C(C(=C1)[N+]([O-])=O)[N+]([O-])=O
1
–
2
of
2
results
1
3,4-Dinitrobenzoic acid, 98+%
CAS: 528-45-0 Molecular Formula: C7H4N2O6 Molecular Weight (g/mol): 212.12 MDL Number: MFCD00007255 InChI Key: OMVRRHJJQILIJX-UHFFFAOYSA-N Synonym: 3,4-dinitrobenzoic acid,benzoic acid, 3,4-dinitro,unii-0l3n4t0gpd,ccris 3128,3,4-dinitrobenzioc acid,0l3n4t0gpd,3,4-dinitrobenzoate,3,4-di-nitro-benzoic acid,zlchem 522,pubchem4956 PubChem CID: 10709 IUPAC Name: 3,4-dinitrobenzoic acid SMILES: OC(=O)C1=CC=C(C(=C1)[N+]([O-])=O)[N+]([O-])=O