CAS RN 528-75-6
IUPAC Name:
2,4-dinitrobenzaldehyde
Synonyms:
2,4-dinitrobenzaldehyde
Molecular Weight (g/mol):
196.12
Molecular Formula:
C7H4N2O5
InChi Key:
ZILXIZUBLXVYPI-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(C=O)C(=C1)[N+]([O-])=O
1
–
2
of
2
results
1
2,4-Dinitrobenzaldehyde, 98%
CAS: 528-75-6 Molecular Formula: C7H4N2O5 Molecular Weight (g/mol): 196.118 MDL Number: MFCD00013376 InChI Key: ZILXIZUBLXVYPI-UHFFFAOYSA-N Synonym: benzaldehyde, 2,4-dinitro,unii-zac919yu3z,ccris 2684,zac919yu3z,pubchem8223,benzaldehyde,4-dinitro,2,4-dinitro-benzaldehyde,acmc-209l2f,3-07-00-00923 beilstein handbook reference,ksc490k9l PubChem CID: 68250 IUPAC Name: 2,4-dinitrobenzaldehyde SMILES: C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C=O
2
2,4-Dinitrobenzaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.