accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 529-36-2

IUPAC Name: (naphthalen-1-yl)sulfanide
Synonyms: naphthalen-1-ylsulfanide
Molecular Weight (g/mol): 159.23
Molecular Formula: C10H7S
InChi Key: SEXOVMIIVBKGGM-UHFFFAOYSA-M
SMILES: [S-]C1=C2C=CC=CC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)