accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOHNNO(E)

CAS RN 5290-62-0

IUPAC Name: (4E)-4-[2-(4-nitrophenyl)hydrazin-1-ylidene]-1,4-dihydronaphthalen-1-one
Synonyms: (4E)-4-[2-(4-nitrophenyl)hydrazin-1-ylidene]naphthalen-1-one
Molecular Weight (g/mol): 293.28
Molecular Formula: C16H11N3O3
InChi Key: UYDXOFSMBKSLPM-OBGWFSINSA-N
SMILES: [O-][N+](=O)C1=CC=C(N\N=C2/C=CC(=O)C3=CC=CC=C23)C=C1
Quantity 
  • (2)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results