CAS RN 52927-22-7
IUPAC Name:
6-hydroxynaphthalene-2-carbonitrile
Synonyms:
6-hydroxynaphthalene-2-carbonitrile
Molecular Weight (g/mol):
169.18
Molecular Formula:
C11H7NO
InChi Key:
WKTNIBWKHNIPQR-UHFFFAOYSA-N
SMILES:
OC1=CC2=CC=C(C=C2C=C1)C#N
1
–
1
of
1
results
1
6-Cyano-2-naphthol, TRC
CAS: 52927-22-7 Molecular Formula: C11 H7 N O Molecular Weight (g/mol): 169.18 IUPAC Name: 6-hydroxynaphthalene-2-carbonitrile SMILES: Oc1ccc2cc(ccc2c1)C#N