accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOCl

CAS RN 5293-97-0

IUPAC Name: bis(2-chlorophenyl)methanone
Synonyms: bis(2-chlorophenyl)methanone
Molecular Weight (g/mol): 251.11
Molecular Formula: C13H8Cl2O
InChi Key: DRDRZHJTTDSOPK-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C(=O)C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results