CAS RN 5297-17-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Succinylmonocholine Chloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
{2-[(3-Carboxypropanoyl)oxy]ethyl}trimethylazanium Chloride, TRC
CAS: 5297-17-6 Molecular Formula: C9 H18 N O4 . Cl Molecular Weight (g/mol): 239.7 Synonym: (3-carboxy-1-oxopropoxy)-N,N,N-trimethyl-ethanaminium chloride (1:1), Succinylmonocholine Chloride, Choline, chloride, hydrogen succinate, Succinic acid, monoester with choline chloride,Succinyl monocholine chloride IUPAC Name: 2-(3-carboxypropanoyloxy)ethyl-trimethylazanium;chloride SMILES: [Cl-].C[N+](C)(C)CCOC(=O)CCC(=O)O