CAS RN 53-86-1
Indomethacin, 98%
CAS: 53-86-1 Molecular Formula: C19H16ClNO4 Molecular Weight (g/mol): 357.79 MDL Number: MFCD00057095 InChI Key: CGIGDMFJXJATDK-UHFFFAOYSA-N Synonym: indomethacin,indometacin,indocin,indometacine,indomethacine,indocid,metindol,amuno,indomethazine,imbrilon PubChem CID: 3715 ChEBI: CHEBI:49662 IUPAC Name: 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid SMILES: CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)O
Thermo Scientific Chemicals Indomethacin
CAS: 53-86-1 Molecular Formula: C19H16ClNO4 Molecular Weight (g/mol): 357.79 InChI Key: CGIGDMFJXJATDK-UHFFFAOYSA-N Synonym: indomethacin,indometacin,indocin,indometacine,indomethacine,indocid,metindol,amuno,indomethazine,imbrilon PubChem CID: 3715 ChEBI: CHEBI:49662 IUPAC Name: 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid SMILES: CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)O
Indomethacin, 99+%
CAS: 53-86-1 Molecular Formula: C19H16ClNO4 Molecular Weight (g/mol): 357.79 MDL Number: MFCD00057095 InChI Key: CGIGDMFJXJATDK-UHFFFAOYSA-N Synonym: indomethacin,indometacin,indocin,indometacine,indomethacine,indocid,metindol,amuno,indomethazine,imbrilon PubChem CID: 3715 ChEBI: CHEBI:49662 IUPAC Name: 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid SMILES: CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)O
Indomethacin, TRC
CAS: 53-86-1 Molecular Formula: C19 H16 Cl N O4 Molecular Weight (g/mol): 357.79 Synonym: 1H-Indole-3-acetic acid, 1-(4-chlorobenzoyl)-5-methoxy-2-methyl- (9CI, ACI),1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic acid (ACI),Indole-3-acetic acid, 1-(p-chlorobenzoyl)-5-methoxy-2-methyl- (8CI),1-(4-Chlorobenzoyl)-2-methyl-5-methoxyindole-3-acetic acid,1-(4-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic acid,1-(p-Chlorobenzoyl)-2-methyl-5-methoxy-3-indolylacetic acid,1-(p-Chlorobenzoyl)-2-methyl-5-methoxyindole-3-acetic acid,1-(p-Chlorobenzoyl)-5-methoxy-2-methyl-3-indolylacetic acid,1-(p-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic acid,2-[1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetic acid,2-[1-(4-Chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid,2-[1-[(4-Chlorophenyl)carbonyl]-5-methoxy-2-methyl-1H-indol-3-yl]acetic acid,[1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetic acid,Aconip,Amuno,Argun,Artracin,Artrinovo,Artrivia,Bonidin,Bonidon Gel,Catlep,Chibro-Amuno,Chrono-Indicid,Chrono-Indocid 75,Confortid,Dolcidium,Dolcidium PL,Dolovin,Durametacin,Elmetacin,Endol,Flam,Globa,Idomethine,Imbrilon,IMC,Inacid,Indacin,Indo-Phlogont,Indo-Rectolmin,Indo-Tablinen,Indocid,Indocid (pharmaceutical),Indocin,Indocin IV,Indocine,Indocollyre,Indomecol,Indomed,Indomee,Indometacin,Indometacine,Indomethacin,Indomethacine,Indomethine,Indomod,Indonol,Indoptic,Indoptol,Indorektal,IndoRich,Indoxen,Indren,Inflazon,Infrocin,Innamit,Inteban,Inteban SP,Lausit,MeSH ID: D007213,Metacen,Metartril,Methazine,Metindol,Mezolin,Microcid,Mikametan,Mobilan,N-(p-Chlorobenzoyl)-2-methyl-5-methoxy-3-indolylacetic acid,NSC 77541,Reumacide,Rheumacin LA,Sadoreum,Tannex,Vital Vitacid,Vonum,α-[1-(p-Chlorobenzoyl)-2-methyl-5-methoxy-3-indolyl]acetic acid IUPAC Name: 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid SMILES: COc1ccc2c(c1)c(CC(=O)O)c(C)n2C(=O)c3ccc(Cl)cc3
Indometacin 1.0 mg/ml in Acetonitrile, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Indometacin, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Indometacin, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Indomethacin, MP Biomedicals™
CAS: 53-86-1 Molecular Formula: C19H16ClNO4 Molecular Weight (g/mol): 357.79 MDL Number: MFCD00057095 InChI Key: CGIGDMFJXJATDK-UHFFFAOYSA-N Synonym: indomethacin,indometacin,indocin,indometacine,indomethacine,indocid,metindol,amuno,indomethazine,imbrilon PubChem CID: 3715 ChEBI: CHEBI:49662 IUPAC Name: 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid SMILES: CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)O