accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3O

CAS RN 530-44-9

IUPAC Name: 4-benzoyl-N,N-dimethylaniline
Synonyms: 4-benzoyl-N,N-dimethylaniline
Molecular Weight (g/mol): 225.29
Molecular Formula: C15H15NO
InChi Key: BEUGBYXJXMVRFO-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results