accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 530-59-6

IUPAC Name: (2E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
Synonyms: sinapinate sinapic acid sinapinic acid
Molecular Weight (g/mol): 224.21
Molecular Formula: C11H12O5
InChi Key: PCMORTLOPMLEFB-ONEGZZNKSA-N
SMILES: COC1=CC(\C=C\C(O)=O)=CC(OC)=C1O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results