accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONNNN

CAS RN 530-62-1

IUPAC Name: 1-(1H-imidazole-1-carbonyl)-1H-imidazole
Synonyms: carbonyldiimidazole
Molecular Weight (g/mol): 162.15
Molecular Formula: C7H6N4O
InChi Key: PFKFTWBEEFSNDU-UHFFFAOYSA-N
SMILES: O=C(N1C=CN=C1)N1C=CN=C1
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (7)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)