accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HO

CAS RN 530-91-6

IUPAC Name: 1,2,3,4-tetrahydronaphthalen-2-ol
Synonyms: 1,2,3,4-tetrahydronaphthalen-2-ol
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: JWQYZECMEPOAPF-UHFFFAOYNA-N
SMILES: OC1CCC2=CC=CC=C2C1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results