accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONOO

CAS RN 53055-05-3

IUPAC Name: 3-methoxy-2-nitrobenzaldehyde
Synonyms: 3-methoxy-2-nitrobenzaldehyde
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: GDTUACILWWLIJF-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C=O)=C1[N+]([O-])=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results