accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OBr

CAS RN 53087-13-1

IUPAC Name: 1-(benzyloxy)-3-bromobenzene
Synonyms: 1-(benzyloxy)-3-bromobenzene
Molecular Weight (g/mol): 263.13
Molecular Formula: C13H11BrO
InChi Key: HVWZMGZBJCJDOX-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(OCC2=CC=CC=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results