accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClCH3NH3CNNH2S

CAS RN 531-53-3

IUPAC Name: 3-amino-7-(dimethylamino)-5λ⁴-phenothiazin-5-ylium chloride
Synonyms: azure A
Molecular Weight (g/mol): 291.8
Molecular Formula: C14H14ClN3S
InChi Key: PGWTYMLATMNCCZ-UHFFFAOYSA-M
SMILES: [Cl-].CN(C)C1=CC=C2N=C3C=CC(N)=CC3=[S+]C2=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results