accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOO

CAS RN 531-59-9

IUPAC Name: 7-methoxy-2H-chromen-2-one
Synonyms: methylumbelliferone herniarin COUMARIN7methoxy herniarin (6CI) ayapanin herniarine methoxycourmarin, 7- 7-methoxycourmarin
Molecular Weight (g/mol): 176.17
Molecular Formula: C10H8O3
InChi Key: LIIALPBMIOVAHH-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CC(=O)OC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)