accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 531-81-7

IUPAC Name: 2-oxo-2H-chromene-3-carboxylic acid
Synonyms: 2-oxochromene-3-carboxylic acid
Molecular Weight (g/mol): 190.15
Molecular Formula: C10H6O4
InChi Key: ACMLKANOGIVEPB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC=CC=C2OC1=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results