accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SiOH3COH3COH3C

CAS RN 5314-55-6

IUPAC Name: ethyltrimethoxysilane
Synonyms: ethyl trimethoxy silane
Molecular Weight (g/mol): 150.25
Molecular Formula: C5H14O3Si
InChi Key: SBRXLTRZCJVAPH-UHFFFAOYSA-N
SMILES: CC[Si](OC)(OC)OC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results