accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HONNH

CAS RN 5317-32-8

IUPAC Name: 3-(piperazin-1-yl)propan-1-ol
Synonyms: 3-(piperazin-1-yl)propan-1-ol
Molecular Weight (g/mol): 144.22
Molecular Formula: C7H16N2O
InChi Key: LWEOFVINMVZGAS-UHFFFAOYSA-N
SMILES: OCCCN1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results