accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 53174-98-4

IUPAC Name: thieno[2,3-b]pyridine-2-carbaldehyde
Synonyms: thieno[2,3-b]pyridine-2-carbaldehyde
Molecular Weight (g/mol): 163.19
Molecular Formula: C8H5NOS
InChi Key: HGZQKYOLRLVMHZ-UHFFFAOYSA-N
SMILES: O=CC1=CC2=CC=CN=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results