accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 532-24-1

IUPAC Name: 8-methyl-8-azabicyclo[3.2.1]octan-3-one
Synonyms: tropanone tropanon 3-tropanone tropan-3-one
Molecular Weight (g/mol): 139.2
Molecular Formula: C8H13NO
InChi Key: QQXLDOJGLXJCSE-UHFFFAOYNA-N
SMILES: CN1C2CCC1CC(=O)C2
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)