CAS RN 532-65-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(+)-ar-Turmerone, TRC
CAS: 532-65-0 Molecular Formula: C15 H20 O Molecular Weight (g/mol): 216.32 Synonym: 2-Hepten-4-one, 2-methyl-6-(4-methylphenyl)-, (6S)-,2-Hepten-4-one, 2-methyl-6-(4-methylphenyl)-, (S)-,2-Hepten-4-one, 2-methyl-6-p-tolyl- (8CI),ar-Turmerone (6CI),(+)-(S)-ar-Turmerone,(+)-Turmerone,(+)-ar-Turmerone,(6S)-2-Methyl-6-(4-methylphenyl)-2-hepten-4-one,(S)-(+)-ar-Turmerone,(S)-ar-Turmerone,ar-(+)-Turmerone,ar-Tumerone IUPAC Name: (6S)-2-methyl-6-(4-methylphenyl)hept-2-en-4-one SMILES: C[C@@H](CC(=O)C=C(C)C)c1ccc(C)cc1