accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNH

CAS RN 5321-49-3

IUPAC Name: 1-(2-phenylethyl)piperazine
Synonyms: 1-(2-phenylethyl)piperazine
Molecular Weight (g/mol): 190.29
Molecular Formula: C12H18N2
InChi Key: LKUAPSRIYZLAAO-UHFFFAOYSA-N
SMILES: C(CC1=CC=CC=C1)N1CCNCC1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)